Thermo Scientific Chemicals

6-Chloropurine 2'-deoxyriboside, 97%, Thermo Scientific Chemicals

Catalog number: J64871.03
1 g, Each
Thermo Scientific Chemicals

6-Chloropurine 2'-deoxyriboside, 97%, Thermo Scientific Chemicals

Catalog number: J64871.03
1 g, Each
Quantity
Catalog number: J64871.03
also known as J64871-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4594-45-0
IUPAC Name
5-(6-chloro-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Formula
C10H11ClN4O3
InChI Key
PGEULCIODBNODW-UHFFFAOYNA-N
SMILES
OCC1OC(CC1O)N1C=NC2=C(Cl)N=CN=C12
Water Content
≤6.0%
Purity
≥96.0%
Form
Powder
Appearance (Color)
White to off-white

Description

6-Chloropurine 2'-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropurine 2′-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

Notes
Light sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text