6-Chloropurine 2'-deoxyriboside, 97%, Thermo Scientific Chemicals
6-Chloropurine 2'-deoxyriboside, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chloropurine 2'-deoxyriboside, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number J64871.MD
also known as J64871-MD
Price (USD)/ Each
97.50
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
97.50
Add to cart
6-Chloropurine 2'-deoxyriboside, 97%, Thermo Scientific Chemicals
Catalog numberJ64871.MD
Price (USD)/ Each
97.50
-
Add to cart
Chemical Identifiers
CAS4594-45-0
IUPAC Name5-(6-chloro-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular FormulaC10H11ClN4O3
InChI KeyPGEULCIODBNODW-UHFFFAOYNA-N
SMILESOCC1OC(CC1O)N1C=NC2=C(Cl)N=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content≤6.0%
Purity≥96.0%
Appearance (Color)White to off-white
FormPowder
6-Chloropurine 2'-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropurine 2′-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

Notes
Light sensitive.
RUO – Research Use Only

General References:

  1. Tanaka, K.; Tainaka, K.; Umemoto, T.; Nomura, A.; Okamoto, A. An Osmium-DNA Interstrand Complex: Application to Facile DNA Methylation Analysis. J. Am. Chem. Soc. 2007, 129 (46), 14511-145.
  2. Rivero, C. W.; Benedetti, E. C. D.; Sambeth, J. E.; Lozano, M. E.; Trelles, J. A. Biosynthesis of anti-HCV compounds using thermophilic microorganisms. Bioorg. Med. Chem. Lett. 2012, 22 (19), 6059-6062.