(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, Thermo Scientific Chemicals
(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 mg
Catalog number J64884.MA
also known as J64884-MA
Price (USD)/ Each
293.00
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
293.00
Add to cart
(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, Thermo Scientific Chemicals
Catalog numberJ64884.MA
Price (USD)/ Each
293.00
-
Add to cart
Specifications
Chemical Name or Material(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
CAS74341-63-2
ColorWhite
Recommended StorageStore at -20°C
TSCANo
View more
Selective quisqualate agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only