Acetylene-PEG4-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals
Acetylene-PEG4-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetylene-PEG4-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals

A carboxyrhodamine 110 based labeling reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
5 mg
Catalog number J64892.MB
also known as J64892-MB
Price (USD)/ Each
885.00
-
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)/ Each
885.00
Add to cart
Acetylene-PEG4-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals
Catalog numberJ64892.MB
Price (USD)/ Each
885.00
-
Add to cart
Specifications
Chemical Name or MaterialAcetylene-PEG4-carboxyrhodamine 110 conjugate
CAS1615739-95-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
A carboxyrhodamine 110 based labeling reagent; reacts with azides via a copper-catalyzed click reactions. Carboxyrhodamine 110 is a bright and photostable green fluorescent label that can be excited at a wavelength of 488nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only