Thermo Scientific Chemicals

N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride

Catalog number: J64915.MD
250 mg, Each
Thermo Scientific Chemicals

N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride

Catalog number: J64915.MD
250 mg, Each
Quantity
Catalog number: J64915.MD
also known as J64915-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21653-40-7
IUPAC Name
hydrogen 5-[(diaminomethylidene)amino]-N-(4-nitrophenyl)-2-(phenylformamido)pentanamide chloride
Molecular Formula
C19H23ClN6O4
InChI Key
DEOKFPFLXFNAON-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Form
Solid
Assay (HPLC)
≥95%
Proton NMR
Conforms to structure

Description

N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.

Solubility
Soluble in DMSO at 50mg/ml

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text