2-Aminopurine, 98%, Thermo Scientific Chemicals
2-Aminopurine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminopurine, 98%, Thermo Scientific Chemicals

A protein kinase R inhibitor
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
250 mg
1 g
Catalog number J64919.ME
also known as J64919-ME
Price (USD)/ Each
315.00
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
315.00
Add to cart
2-Aminopurine, 98%, Thermo Scientific Chemicals
Catalog numberJ64919.ME
Price (USD)/ Each
315.00
-
Add to cart
Specifications
Chemical Name or Material2-Aminopurine
CAS452-06-2
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
A protein kinase R inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only