Thermo Scientific Chemicals

2'-O-Methyladenosine, 99%

Catalog number: J64922.03
1 g, Each
Thermo Scientific Chemicals

2'-O-Methyladenosine, 99%

Catalog number: J64922.03
1 g, Each
Quantity
Catalog number: J64922.03
also known as J64922-03
Price (USD)
Quantity
-
Have Questions?

Description

2'-O-Methyladenosine is an analog of adenosine used to prepare nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-O-Methyladenosine is an analog of adenosine used to prepare nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

Solubility
Soluble in water.

Notes
Store away from st
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text