Thermo Scientific Chemicals

O-Acetyl-L-serine hydrochloride

Catalog number: J64933.06
5 g, Each
Thermo Scientific Chemicals

O-Acetyl-L-serine hydrochloride

Catalog number: J64933.06
5 g, Each
Quantity
Catalog number: J64933.06
also known as J64933-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66638-22-0
IUPAC Name
3-(acetyloxy)-2-aminopropanoic acid
Molecular Formula
C5H9NO4
InChI Key
VZXPDPZARILFQX-UHFFFAOYNA-N
SMILES
CC(=O)OCC(N)C(O)=O
Assay from Supplier's CofA
≥96.0%
Carbon-13 NMR
Conforms to structure
Appearance (Color)
White
Proton NMR
Conforms to structure
Form
Powder

Description

O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.

Solubility
Soluble in ethanol almost transparency.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text