Thermo Scientific Chemicals

2'-Deoxythymidine-5'-diphosphate trisodium salt, 97%

Catalog number: J64938.MB
25 mg, Each
Thermo Scientific Chemicals

2'-Deoxythymidine-5'-diphosphate trisodium salt, 97%

Catalog number: J64938.MB
25 mg, Each
Quantity
Catalog number: J64938.MB
also known as J64938-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95648-78-5
IUPAC Name
trisodium ({[(2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular Formula
C10H13N2Na3O11P2
InChI Key
HBYPEEZBYHQLDM-SPSULGLQSA-K
SMILES
[Na+].[Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])([O-])=O)O2)C(=O)NC1=O
Appearance (Color)
White
Form
Powder
Assay from Suppliers CofA
≥95.0% (HPLC)
Water Content
≤11.0%

Description

2'-Deoxythymidine-5'-diphosphate trisodium salt is used as a stable nucleotide useful for PCR and RT-PCR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxythymidine-5′-diphosphate trisodium salt is used as a stable nucleotide useful for PCR and RT-PCR.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text