MIF Antagonist, ISO-1
MIF Antagonist, ISO-1
Thermo Scientific Chemicals

MIF Antagonist, ISO-1

A cell-permeable isoxazoline compound
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number J64947.LB0
also known as J64947-#0
Price (USD)/ Each
118.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
118.00
Add to cart
MIF Antagonist, ISO-1
Catalog numberJ64947.LB0
Price (USD)/ Each
118.00
-
Add to cart
Chemical Identifiers
CAS478336-92-4
IUPAC Namemethyl 2-[3-(4-oxocyclohexa-2,5-dien-1-ylidene)-1,2-oxazolidin-5-yl]acetate
Molecular FormulaC12H13NO4
InChI KeyCLVQGFPQFNASJH-UHFFFAOYNA-N
SMILESCOC(=O)CC1CC(NO1)=C1C=CC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay from Supplier's CofA≥95%
FormCrystals or powder or crystalline powder
MIF Antagonist, ISO-1 inhibits MIF dopachrome tautomerase activity by binding to its catalytic active site (IC50 = 7 μM for D-dopachrome tautomerase) and suppresses the production of TNFα, PGE2 and COX-2 in human monocytes and arachidonic acid in RAW 264.7 macrophages. MIF Antagonist, ISO-1 has been shown to exhibit antidiabetogenic properties in immunoinflammatory diabetic mouse model.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C