Thermo Scientific Chemicals

MIF Antagonist, ISO-1

Catalog number: J64947.MB
25 mg, Each
Thermo Scientific Chemicals

MIF Antagonist, ISO-1

Catalog number: J64947.MB
25 mg, Each
Quantity
Catalog number: J64947.MB
also known as J64947-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
478336-92-4
IUPAC Name
methyl 2-[3-(4-oxocyclohexa-2,5-dien-1-ylidene)-1,2-oxazolidin-5-yl]acetate
Molecular Formula
C12H13NO4
InChI Key
CLVQGFPQFNASJH-UHFFFAOYNA-N
SMILES
COC(=O)CC1CC(NO1)=C1C=CC(=O)C=C1
Appearance (Color)
White to pale yellow
Assay from Supplier's CofA
≥95%
Form
Crystals or powder or crystalline powder

Description

MIF Antagonist, ISO-1 inhibits MIF dopachrome tautomerase activity by binding to its catalytic active site (IC50 = 7 μM for D-dopachrome tautomerase) and suppresses the production of TNFα, PGE2 and COX-2 in human monocytes and arachidonic acid in RAW 264.7 macrophages. MIF Antagonist, ISO-1 has been shown to exhibit antidiabetogenic properties in immunoinflammatory diabetic mouse model.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text