Thermo Scientific Chemicals

Azido-PEG3-biotin conjugate, Thermo Scientific Chemicals

Catalog number: J64996.MC
100 mg, Each
Thermo Scientific Chemicals

Azido-PEG3-biotin conjugate, Thermo Scientific Chemicals

Catalog number: J64996.MC
100 mg, Each
Quantity
Catalog number: J64996.MC
also known as J64996-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
875770-34-6
IUPAC Name
N-(2-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}ethyl)-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
Molecular Formula
C18H32N6O5S
InChI Key
ZWFOOMQCYIGZBE-UHFFFAOYNA-N
SMILES
[N-]=[N+]=NCCOCCOCCOCCNC(=O)CCCCC1SCC2NC(=O)NC12
Form
Amorphous solid
LCMS
Conforms
Appearance (Color)
White
Assay from Supplier's CofA
≥90%

Description

Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it?s components do not react with the biological environment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it′s components do not react with the biological environment.

Solubility
Solubile in DMSO, DMF and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text