Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Albumin, Bovine, Fraction V, 35% Solution, Reagent Grade, Fatty Acid Free, No Preservative, Thermo Scientific Chemicals

Catalog number: J65042.AE
100mL, Each
Thermo Scientific Chemicals

Albumin, Bovine, Fraction V, 35% Solution, Reagent Grade, Fatty Acid Free, No Preservative, Thermo Scientific Chemicals

Catalog number: J65042.AE
100mL, Each
Quantity
Catalog number: J65042.AE
also known as J65042-AE
Price (USD)
Have Questions?
Form
Liquid
Appearance (Color)
Clear, yellow to green
Assay from Supplier's CofA
34.0-36.0 g/dL
Comment
Preservative: N/A (none present)
pH
6.6-8.5

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text