It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.
Solubility
Soluble in DMSO, DMF or methanol.
Notes
Excite using the 488nm spectral line of the argon ion laser
RUO – Research Use Only
General References:
- Yong Liang; Xin Jiang; Nannan Tang; Limin Yang; Haifeng Chen; Qiuquan Wang. Quantification and visualization of glutathione S-transferase μ 1 in cells using inductively coupled plasma mass spectrometry (ICP-MS) and fluorescence microscopy. Analytical and Bioanalytical Chemistry. 2015, 407, (9), 2373-2381.
- Sarah A. Sabatinos; Susan L. Forsburg.Measuring DNA Content by Flow Cytometry in Fission Yeast. DNA Replication. 1972, 1300, 79-97.