Thermo Scientific Chemicals

1-Amino-1-cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: J65118.03
1 g, Each
Thermo Scientific Chemicals

1-Amino-1-cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: J65118.03
1 g, Each
Quantity
Catalog number: J65118.03
also known as J65118-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22059-21-8
IUPAC Name
1-aminocyclopropane-1-carboxylic acid
Molecular Formula
C4H7NO2
InChI Key
PAJPWUMXBYXFCZ-UHFFFAOYSA-N
SMILES
NC1(CC1)C(O)=O
Assay (unspecified)
>97.5%

Description

1-Aminocyclopropane-1-carboxylic Acid is an intermediate in the conversion of methionine to ethylene during the ripening in apples. It also plays an important role in the biosynthesis of the plant hormone ethylene.1-Aminocyclopropane-1-carboxylic Acid is shown to block convulsions and death produced by NMDA exposure, significantly reducing seizure induction and cell death of NMDA-treated hippocampal neurons. 1-Aminocyclopropane-1-carboxylic Acid is produced endogenously in the tomato and other higher plants as a product of the action of 1-aminocyclopropane-1-carboxylic acid synthase in the biosynthesis of ethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminocyclopropane-1-carboxylic Acid is an intermediate in the conversion of methionine to ethylene during the ripening in apples. It also plays an important role in the biosynthesis of the plant hormone ethylene.1-Aminocyclopropane-1-carboxylic Acid is shown to block convulsions and death produced by NMDA exposure, significantly reducing seizure induction and cell death of NMDA-treated hippocampal neurons. 1-Aminocyclopropane-1-carboxylic Acid is produced endogenously in the tomato and other higher plants as a product of the action of 1-aminocyclopropane-1-carboxylic acid synthase in the biosynthesis of ethylene.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text