Thermo Scientific Chemicals

3'-Azido-3'-deoxythymidine, 98%

Catalog number: J65127.03
1 g, Each
Thermo Scientific Chemicals

3'-Azido-3'-deoxythymidine, 98%

Catalog number: J65127.03
1 g, Each
Quantity
Catalog number: J65127.03
also known as J65127-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30516-87-1
IUPAC Name
1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C10H13N5O4
InChI Key
HBOMLICNUCNMMY-CFQLRCIDNA-N
SMILES
CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O
Appearance (Color)
White to yellow
Form
Powder
Assay from Supplier's CofA
≥97.5%
Infrared spectrum
Conforms

Description

3'-Azido-3'-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3?-Azido-3?-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Azido-3′-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3′-Azido-3′-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text