Thermo Scientific Chemicals

Coenzyme Q-10, 98+%, Thermo Scientific Chemicals

Catalog number: J65137.06
5 g, Each
Thermo Scientific Chemicals

Coenzyme Q-10, 98+%, Thermo Scientific Chemicals

Catalog number: J65137.06
5 g, Each
Quantity
Catalog number: J65137.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
303-98-0
IUPAC Name
2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaen-1-yl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Molecular Formula
C59H90O4
InChI Key
ACTIUHUUMQJHFO-UPTCCGCDSA-N
SMILES
COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O
Assay (unspecified)
>98.0%
Residue on ignition
<0.1%
Loss on Drying
0.04%

Description

It is used as a component of electron transport chain and in aerobic cellular respiration, Coenzyme Q10 can protect human endothelial cells from oxidative damage by modulating the nitric oxide pathway. In addition, Coenzyme Q10 inhibits LDL oxidation, a major process in the development of atherosclerosis. Coenzyme Q10 is known to suppress the down regulation of NOS and up-regulation of NOS2. As a result of its ability to bind Ca2+, Coenzyme Q10 can transport this cation across artificial biomimetic membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a component of electron transport chain and in aerobic cellular respiration, Coenzyme Q10 can protect human endothelial cells from oxidative damage by modulating the nitric oxide pathway. In addition, Coenzyme Q10 inhibits LDL oxidation, a major process in the development of atherosclerosis. Coenzyme Q10 is known to suppress the down regulation of NOS and up-regulation of NOS2. As a result of its ability to bind Ca2+, Coenzyme Q10 can transport this cation across artificial biomimetic membranes.

Solubility
Soluble in chloroform.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text