4-Amino-3-hydroxybutyric acid, Thermo Scientific Chemicals
4-Amino-3-hydroxybutyric acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3-hydroxybutyric acid, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number J65145.06
also known as J65145-06
Price (USD)/ Each
64.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
64.80
Add to cart
4-Amino-3-hydroxybutyric acid, Thermo Scientific Chemicals
Catalog numberJ65145.06
Price (USD)/ Each
64.80
-
Add to cart
Specifications
Chemical Name or Material4-Amino-3-hydroxybutyric acid
Melting Point223°C (decomposition)
CAS924-49-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Reduces uptake of iodine by the thyroid glands in animals. Antispasmodic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Reduces uptake of iodine by the thyroid glands in animals
  • Antispasmodic
RUO – Research Use Only