Thermo Scientific Chemicals

Chemotactic Peptide

Catalog number: J65184.MB
25 mg, Each
Thermo Scientific Chemicals

Chemotactic Peptide

Catalog number: J65184.MB
25 mg, Each
Quantity
Catalog number: J65184.MB
also known as J65184-MB
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Off-white
Form
Lyophilized Powder
Assay (special)
Peptide content: ≥90%
Mass Spec
Conforms
Purity
≥99.0%

Description

Induces leucocyte chemotaxis, and activates macrophages

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text