Thermo Scientific Chemicals

2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate, 98%

Catalog number: J65186.03
1 g, Each
Thermo Scientific Chemicals

2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate, 98%

Catalog number: J65186.03
1 g, Each
Quantity
Catalog number: J65186.03
also known as J65186-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
146877-98-7
Appearance (Color)
White
Purity
≥97.0% (HPLC)
Form
Powder
Water Content
≤20.0%

Description

2'-Deoxyguanosine-5'-monophosphate Disodium Salt Hydrate is a reactant involved in analysis of self-assembling in solution and nucleation/growth of G-qudruplexes, Nucleophilic trapping, Reductive alkylation. It is a nucleotide precursor used in DNA synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyguanosine-5′-monophosphate Disodium Salt Hydrate is a reactant involved in analysis of self-assembling in solution and nucleation/growth of G-qudruplexes, Nucleophilic trapping, Reductive alkylation. It is a nucleotide precursor used in DNA synthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text