Thermo Scientific Chemicals
N-Acetyl-Asp-Glu-Val-Asp-al
Catalog number: J65204.LB0
5 mg, Each
Thermo Scientific Chemicals
N-Acetyl-Asp-Glu-Val-Asp-al
Catalog number: J65204.LB0
5 mg, Each
Quantity
Catalog number: J65204.LB0
also known as J65204-#0
Price (USD)
Have Questions?
Description
Reversible inhibitor of interleukin-1beta converting enzyme (ICE); also inhibits caspase-3 cleavage of poly(ADP-ribose) polymerase