Thermo Scientific Chemicals

Bromocresol Green, ACS

Catalog number: J65225.06
5 g, Each
Thermo Scientific Chemicals

Bromocresol Green, ACS

Catalog number: J65225.06
5 g, Each
Quantity
Catalog number: J65225.06
also known as J65225-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-60-8
IUPAC Name
3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Formula
C21H14Br4O5S
InChI Key
FRPHFZCDPYBUAU-UHFFFAOYSA-N
SMILES
CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C
Comment
Clarity of solution P.T.
Comment
Visual transition interval pH 3.8 (yellow) to pH 5.4 (blue)

Description

Bromocresol Green is used in a variety of colorimetric detection technologies. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes. In TLC, it is used for visualization of the compounds with functional groups whose pKa is below 5.0. And it is ready-to-use spray and dip reagent for chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol Green is used in a variety of colorimetric detection technologies. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes. In TLC, it is used for visualization of the compounds with functional groups whose pKa is below 5.0. And it is ready-to-use spray and dip reagent for chromatography.

Solubility
Soluble in water, ethanol, methyl cellosolve.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text