Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Biliverdin hydrochloride, Thermo Scientific Chemicals

Catalog number: J65226.MD
250 mg, Each
Thermo Scientific Chemicals

Biliverdin hydrochloride, Thermo Scientific Chemicals

Catalog number: J65226.MD
250 mg, Each
Quantity
Catalog number: J65226.MD
also known as J65226-MD
Price (USD)
Have Questions?

Chemical Identifiers

CAS
55482-27-4
IUPAC Name
dihydrogen 3-(2-{[(2Z,5E)-3-(2-carboxyethyl)-5-[(3-ethenyl-4-methyl-2-oxo-2H-pyrrol-5-yl)methylidene]-4-methyl-2,5-dihydro-1H-pyrrol-2-ylidene]methyl}-5-{[(2Z)-3-ethenyl-4-methyl-5-oxo-2,5-dihydro-1H-pyrrol-2-ylidene]methyl}-4-methyl-1H-pyrrol-3-yl)propanoic acid dichloride
Molecular Formula
C33H36Cl2N4O6
InChI Key
GNLHFBAZSKVVCQ-DKUYZEAMSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].CC1=C(C=C)\C(NC1=O)=C\C1=C(C)C(CCC(O)=O)=C(N1)\C=C1/N\C(=C\C2=NC(=O)C(C=C)=C2C)C(C)=C1CCC(O)=O
Form
Solid
Assay from Suppliers CofA
≥95.0%
Appearance (Color)
Green

Description

Biliverdin dihydrochloride was found responsible for cytoprotection. It was also found to induce tolerance to cardiac allografts. Biliverdin hydrochloride may regulate the cellular heme degradation process by occupying the heme binding site on heme oxygenase which would prevent access of the substrate to the catalytic site of the enzyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biliverdin dihydrochloride was found responsible for cytoprotection. It was also found to induce tolerance to cardiac allografts. Biliverdin hydrochloride may regulate the cellular heme degradation process by occupying the heme binding site on heme oxygenase which would prevent access of the substrate to the catalytic site of the enzyme.

Solubility
Soluble in DMSO, DMF, and basic aqueous solutions

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text