Orexin B, Mouse, Rat
Orexin B, Mouse, Rat
Thermo Scientific Chemicals

Orexin B, Mouse, Rat

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J65275.EXD
also known as J65275-!D
Price (USD)/ Each
225.00
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
225.00
Add to cart
Orexin B, Mouse, Rat
Catalog numberJ65275.EXD
Price (USD)/ Each
225.00
-
Add to cart
Specifications
SpeciesMouse, Rat
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)2935.38
Product TypeOrexin B, mouse, rat
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C