Apoptosis Activator 2, Thermo Scientific Chemicals
Apoptosis Activator 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Apoptosis Activator 2, Thermo Scientific Chemicals
Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines
Have Questions?
Quantity:
5 mg
Catalog number J65320.LB0
also known as J65320-#0
Price (USD)/ Each
95.40
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
95.40
Add to cart
Apoptosis Activator 2, Thermo Scientific Chemicals
Catalog numberJ65320.LB0
Price (USD)/ Each
95.40
-
Add to cart
Specifications
DescriptionOrange
Quantity5 mg
Recommended StorageKeep cold; Store under Argon
CAS79183-19-0
Molecular FormulaC15H9Cl2NO2, C15H9Cl2NO2
View more
Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C