Thermo Scientific Chemicals

Cafestol, Thermo Scientific Chemicals

Catalog number: J65355.MF
50 mg, Each
Thermo Scientific Chemicals

Cafestol, Thermo Scientific Chemicals

Catalog number: J65355.MF
50 mg, Each
Quantity
Catalog number: J65355.MF
also known as J65355-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
469-83-0
IUPAC Name
(1S,4S,12S,13R,16R,17R)-17-(hydroxymethyl)-12-methyl-8-oxapentacyclo[14.2.1.0¹,¹³.0⁴,¹².0⁵,⁹]nonadeca-5(9),6-dien-17-ol
Molecular Formula
C20H28O3
InChI Key
DNJVYWXIDISQRD-HWUKTEKMSA-N
SMILES
C[C@@]12CCC3=C(C=CO3)[C@H]1CC[C@@]13C[C@@H](CC[C@@H]21)[C@@](O)(CO)C3
Appearance (Color)
White to off white
Form
Powder
Assay (HPLC)
≥ 98.0% (UHPLC)
Assay (special)
Identification: Conforms

Description

Natural extract from coffee beans which induces glutathione S-transferase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text