3'-Deoxyadenosine, 98%, Thermo Scientific Chemicals
3'-Deoxyadenosine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3'-Deoxyadenosine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J65366.MB
also known as J65366-MB
Price (USD)/ Each
186.00
-
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)/ Each
186.00
Add to cart
3'-Deoxyadenosine, 98%, Thermo Scientific Chemicals
Catalog numberJ65366.MB
Price (USD)/ Each
186.00
-
Add to cart
Chemical Identifiers
CAS73-03-0
IUPAC Name(2R,3R,5S)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolan-3-ol
Molecular FormulaC10H13N5O3
InChI KeyOFEZSBMBBKLLBJ-PNFQIUPPNA-N
SMILESNC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only