Azadibenzocyclooctyne-maleimide, Thermo Scientific Chemicals
Azadibenzocyclooctyne-maleimide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Azadibenzocyclooctyne-maleimide, Thermo Scientific Chemicals

For incorporation of ADIBO functionality into sulfhydryl containing compounds
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J65377.MB
also known as J65377-MB
Price (USD)/ Each
164.00
-
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)/ Each
164.00
Add to cart
Azadibenzocyclooctyne-maleimide, Thermo Scientific Chemicals
Catalog numberJ65377.MB
Price (USD)/ Each
164.00
-
Add to cart
Specifications
Chemical Name or MaterialAzadibenzocyclooctyne-maleimide
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
ColorWhite to Gray
View more
For incorporation of ADIBO functionality into sulfhydryl containing compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only