Orexin A, Human, Bovine, Rat, Mouse
Orexin A, Human, Bovine, Rat, Mouse
Thermo Scientific Chemicals

Orexin A, Human, Bovine, Rat, Mouse

 
Have Questions?
Quantity:
0.5 mg
Catalog number J65378.EXD
also known as J65378-!D
Price (USD)/ Each
565.00
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
565.00
Add to cart
Orexin A, Human, Bovine, Rat, Mouse
Catalog numberJ65378.EXD
Price (USD)/ Each
565.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
SpeciesBovine, Mouse, Rat
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)3561.09
Product TypeOrexin A, human, bovine, rat, mouse
View more
appetite regulation, wakefulness, locomotor activity, hypothalamic-pituitary-adrenal activity, and pain thresholds. Orexin signaling is activated by nutrient depletion, causing an increase in food intake by delaying the signals of satiety. It activates the orexin-1 and -2 receptors with equal affinity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C