Thermo Scientific Chemicals

Ubiquitin E1 Inhibitor, PYR-41

Catalog number: J65388.MB
25 mg, Each
Thermo Scientific Chemicals

Ubiquitin E1 Inhibitor, PYR-41

Catalog number: J65388.MB
25 mg, Each
Quantity
Catalog number: J65388.MB
also known as J65388-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
418805-02-4
IUPAC Name
ethyl 4-{4-[(5-nitrofuran-2-yl)methylidene]-3,5-dioxopyrazolidin-1-yl}benzoate
Molecular Formula
C17H13N3O7
InChI Key
ARGIPZKQJGFSGQ-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=C(C=C1)N1NC(=O)C(=CC2=CC=C(O2)[N+]([O-])=O)C1=O
Purity
≥97.0%
Appearance (Color)
Dark red to brown
Form
Powder

Description

PYR 41 is an irreversible and cell-permeable UBE1 inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text