Thermo Scientific Chemicals

Artemisinin, 98%, Thermo Scientific Chemicals

Catalog number: J65406.03
1 g, Each
Thermo Scientific Chemicals

Artemisinin, 98%, Thermo Scientific Chemicals

Catalog number: J65406.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J65406.03
also known as J65406-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63968-64-9
IUPAC Name
(1R,4S,5R,8S,9R,12S,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecan-10-one
Molecular Formula
C15H22O5
InChI Key
BLUAFEHZUWYNDE-NNWCWBAJSA-N
SMILES
C[C@@H]1CC[C@H]2[C@@H](C)C(=O)O[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23OO4
Appearance (Color)
White
Infrared spectrum
Conforms
Assay from Supplier's CofA
≥97.5% (HPLC)
Form
Crystals or crystalline powder
Optical Rotation
+70.0 to +80.0°

Description

Artemisinin inhibits angiogenesis by down-regulating HIF-1α and VEGF expression in mouse embryonic stem cells. Artemisinin crosses the blood-brain barrier and is an inhibitor of human NOS2 (iNOS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble to 100mM in DMSO and to 75mM in ethanol

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text