Thermo Scientific Chemicals

2'-Fluoro-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals

Catalog number: J65441.03
1 g, Each
Thermo Scientific Chemicals

2'-Fluoro-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals

Catalog number: J65441.03
1 g, Each
Quantity
Catalog number: J65441.03
also known as J65441-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64183-27-3
IUPAC Name
(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Molecular Formula
C10H12FN5O3
InChI Key
ZGYYPTJWJBEXBC-GPGUAWMRNA-N
SMILES
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3F)C2=NC=N1
Appearance (Color)
White to off-white
Form
Powder
Assay (HPLC)
98.5% min.
Water Content (Karl Fischer Titration)
8.0% max.
Solution Test
10mg/ml in 1M NaOH: clear, colorless solution

Description

2'-Fluoro-2'-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2'-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-2′-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2′-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text