Thermo Scientific Chemicals

5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals

Catalog number: J65443.06
5 g, Each
Thermo Scientific Chemicals

5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals

Catalog number: J65443.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J65443.06
also known as J65443-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30007-47-7
IUPAC Name
5-bromo-5-nitro-1,3-dioxane
Molecular Formula
C4H6BrNO4
InChI Key
XVBRCOKDZVQYAY-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1(Br)COCOC1
Assay (unspecified)
97.5% min.
Melting Point
approx. 60?C
Proton NMR
Conforms to structure

Description

5-Bromo-5-nitro-1,3-dioxane is used as a stabilizer and preserving agent for biological molecules and solutions such as antibodies and antisera. Bronidox is used in a variety of rinse-off cosmetic. It can be used alone or in combination with methylisothiazolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 5-Bromo-5-nitro-1,3-dioxane is a chemical compound showing antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi
• This compound is capable of promoting the oxidation of essential protein thiols, causing inhibition of enzyme activity leading to the inhibition of microbial growth

Application

• 5-Bromo-5-nitro-1,3-dioxane has been used as a stabilizer and preserving agent for biological molecules and solutions, including antibodies and antisera
• It can be used alone or combined with methylisothiazolinone, which is also considered to be an effective preservative

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text