Linomide, Thermo Scientific Chemicals
Linomide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Linomide, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 mg
Catalog number J65453.MA
also known as J65453-MA
Price (USD)/ Each
200.00
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
200.00
Add to cart
Linomide, Thermo Scientific Chemicals
Catalog numberJ65453.MA
Price (USD)/ Each
200.00
-
Add to cart
Specifications
Chemical Name or MaterialLinomide
CAS84088-42-6
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
An immunomodulator with antitumour properties. Inhibits T-cell apoptosis; also inhibits angiogenesis in rats

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An immunomodulator with antitumour properties
  • Inhibits T-cell apoptosis; also inhibits angiogenesis in rats
RUO – Research Use Only