Azido-PEG3-sulforhodamine 101 conjugate, Thermo Scientific Chemicals
Azido-PEG3-sulforhodamine 101 conjugate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Azido-PEG3-sulforhodamine 101 conjugate, Thermo Scientific Chemicals

A water soluble sulforhodamine 101 with azido group functionality
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number J65476.LB0
also known as J65476-#0
Price (USD)/ Each
225.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
225.00
Add to cart
Azido-PEG3-sulforhodamine 101 conjugate, Thermo Scientific Chemicals
Catalog numberJ65476.LB0
Price (USD)/ Each
225.00
-
Add to cart
Specifications
Chemical Name or MaterialAzido-PEG3-sulforhodamine 101 conjugate
CAS1353153-76-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
A water soluble sulforhodamine 101 with azido group functionality

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only