Thermo Scientific Chemicals

2-Deoxy-D-galactose, 99%

Catalog number: J65482.06
5 g, Each
Thermo Scientific Chemicals

2-Deoxy-D-galactose, 99%

Catalog number: J65482.06
5 g, Each
Quantity
Catalog number: J65482.06
also known as J65482-06
Price (USD)
Quantity
-
Have Questions?

Description

2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text