2-Deoxy-D-galactose, 99%
2-Deoxy-D-galactose, 99%
2-Deoxy-D-galactose, 99%
2-Deoxy-D-galactose, 99%
Thermo Scientific Chemicals

2-Deoxy-D-galactose, 99%

For studying galactose uptake into Escherichia coli | CAS: 1949-89-9 | C6H12O5 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
5 g
Catalog number J65482.09
also known as J65482-09
Price (USD)
409.65
Online Exclusive
455.00
Save 45.35 (10%)
Each
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
409.65
Online Exclusive
455.00
Save 45.35 (10%)
Each
Add to cart
Specifications
Beilstein1723333
CAS1949-89-9
ChEBICHEBI:27411
Chemical Name or Material2-Deoxy-D-galactose
ColorWhite
View more
Ask our AI about this Product
2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
RUO – Research Use Only

General References:

  1. Wang, H. Q.; Zhu, L. W.; Hu, X. G.; Chen, N. Dependence of enthalpic pairwise self-interactions on ionic strength: Some 2-deoxy and N-acetyl monosaccharides in aqueous NaCl solutions at T = 298.15 K. J. Chem. Thermodyn. 2015, 90, 8-14.
  2. Sørensen, M.; Frisch, K.; Bender, D.; Keiding, S. The potential use of 2-[18F] fluoro-2-deoxy-D-galactose as a PET/CT tracer for detection of hepatocellular carcinoma. Euro. J. Nucl. Med. Mol. Imaging 2011, 38 (9), 1723-1731.
  3. Krug, M.; Jork, R.; Reymann, K.; Wagner, M.; Matthies, H. The amnesic substance 2-deoxy-D-galactose suppresses the maintenance of hippocampal LTP. Brain Res. 1991, 540 (1-2), 237-242.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link