Thermo Scientific Chemicals

Guanidine hydrochloride, ultrapure, 99%

Catalog number: J65485.A1
1 kg, Each
Thermo Scientific Chemicals

Guanidine hydrochloride, ultrapure, 99%

Catalog number: J65485.A1
1 kg, Each
Quantity
Catalog number: J65485.A1
also known as J65485-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-01-1
IUPAC Name
hydrogen guanidine chloride
Molecular Formula
CH6ClN3
InChI Key
PJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(N)=N
Solution Test
Soluble in water at 6M concentration
RNase
None detected
Assay (special)
≥99.0%
Melting Point
184 to 188°C
Form
Crystals or crystalline powder

Description

Guanidine hydrochloride is used for purification of proteins and nucleic acids. It can be used as a medicine, organic synthesis intermediate and is used in dyes. It acts as an intermediate in the preparation of sulfadiazine, which is an important raw material for sulfamethyldiazine, sulfamethazine and folic acid. It is utilized to synthesis of 2-amino-pyrimidine, 2- amino -6- methyl-pyrimidine and 2-amino -4,6 - dimethyl-pyrimidine. Also used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanidine hydrochloride is used for purification of proteins and nucleic acids. It can be used as a medicine, organic synthesis intermediate and is used in dyes. It acts as an intermediate in the preparation of sulfadiazine, which is an important raw material for sulfamethyldiazine, sulfamethazine and folic acid. It is utilized to synthesis of 2-amino-pyrimidine, 2- amino -6- methyl-pyrimidine and 2-amino -4,6 - dimethyl-pyrimidine. Also used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

Solubility
Soluble in water and ethanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.May agglomerate upon storage.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text