Thermo Scientific Chemicals

2'-Amino-2'-deoxyadenosine, 98%

Catalog number: J65532.03
1 g, Each
Thermo Scientific Chemicals

2'-Amino-2'-deoxyadenosine, 98%

Catalog number: J65532.03
1 g, Each
Quantity
Catalog number: J65532.03
also known as J65532-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10414-81-0
IUPAC Name
(2R,3S,4R,5R)-4-amino-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Formula
C10H14N6O3
InChI Key
CQKMBZHLOYVGHW-GPGUAWMRNA-N
SMILES
N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=CN=C12
Form
Powder
Appearance (Color)
White to pale yellow to pale cream
Assay from Supplier's CofA
≥97.5% (HPLC)

Description

2'-AdA can be used as starting material for the synthesis of 2'-amino nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-AdA can be used as starting material for the synthesis of 2′-amino nucleotides.

Solubility
Soluble in DMSO.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text