Thermo Scientific Chemicals

2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%, Thermo Scientific Chemicals

Catalog number: J65535.06
5 g, Each
Thermo Scientific Chemicals

2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%, Thermo Scientific Chemicals

Catalog number: J65535.06
5 g, Each
Quantity
Catalog number: J65535.06
also known as J65535-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30931-67-0
IUPAC Name
diammonium (2Z)-3-ethyl-2-{2-[(2Z)-3-ethyl-6-sulfonato-2,3-dihydro-1,3-benzothiazol-2-ylidene]hydrazin-1-ylidene}-2,3-dihydro-1,3-benzothiazole-6-sulfonate
Molecular Formula
C18H24N6O6S4
InChI Key
OHDRQQURAXLVGJ-AXMZSLBLSA-N
SMILES
[NH4+].[NH4+].CCN1C(SC2=CC(=CC=C12)S([O-])(=O)=O)=NN=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O
Assay from Supplier's CofA
≥97.5% (HPLC)
Water Content (Karl Fischer Titration)
≤2.0%
Solution Test
Complete: Clear pale green solution (0.5 gm in 1 ml of water)
UV/Vis
Absorbance at 436nm: ≤ 0.25%
UV/Vis
Absorbance at 748nm: ≤ 0.25%

Description

Chromogenic substrate for peroxidase assays.2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromogenic substrate for peroxidase assays.2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.

Solubility
Dissolve in water at 50mg/ml. Solution may be slightly hazy.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text