Albumin, Bovine, Fraction V, 30% Solution, Standard Grade, with Azide as Preservative, Thermo Scientific Chemicals
This product is animal origin and can only be shipped within the United States
Albumin, Bovine, Fraction V, 30% Solution, Standard Grade, with Azide as Preservative, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Albumin, Bovine, Fraction V, 30% Solution, Standard Grade, with Azide as Preservative, Thermo Scientific Chemicals

Albumin bovine/fraction V, CAS # 9048-46-8, also known as bovine serum albumin (BSA), is a serum albumin protein derived from cows.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500mL
100mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65569.AP
also known as J65569-AP
Price (USD)/ Each
450.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
530.00 
Save 79.35 (15%)
-
Add to cart
Quantity:
500mL
Request bulk or custom format
Price (USD)/ Each
450.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
530.00 
Save 79.35 (15%)
Add to cart
Albumin, Bovine, Fraction V, 30% Solution, Standard Grade, with Azide as Preservative, Thermo Scientific Chemicals
Catalog numberJ65569.AP
Price (USD)/ Each
450.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
530.00 
Save 79.35 (15%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Concentration30% w/v
FormulationLiquid
Recommended StorageKeep cold
EINECS Number232-936-2
Quantity500mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Albumin bovine/fraction V is a serum albumin protein commonly used in laboratory experiments including, enzyme-linked immunosorbent assay (ELISAs), immunoblots and immune-histochemistry or -fluorescence
Applications
• Albumin bovine/fraction V displays blocking activity mainly when casein-sensitive antibodies, including phospho-specific antibodies, are used
• In genetic approaches this compound can be used to increase polymerase chain reactions (PCR) yields from low purity templates
• This compound is useful for stabilizing enzymes during storage and also for enzymatic reactions where the absence of nucleases is crucial
• In in vitro cultures, albumin bovine/fraction V has been applied as a nutrient in cell and microbial cultures especially when serum-free media is used
• It is commonly used as a protein standard to obtain calibration curves for several laboratorial experiments
• Albumin bovine/fraction V is a small protein commonly used in immunohistochemistry to block nonspecific regions. The binding of this compound to nonspecific binding sites increases the probability of the antibodies binds only to the region of interest.
RUO – Research Use Only

General References:

  1. Fujiwara, M.; Imura, T. Transparent silica thin films prepared from sodium silicate and bovine serum albumin with petal effect. Ceram. Int. 2015, 41 (6), 7565-7572.
  2. Pereira, M. M.; Pedro, S. N.; Quental, M. V.; Lima, Á. S.; Coutinho, J. A. P.; Freire, M. G. Enhanced extraction of bovine serum albumin with aqueous biphasic systems of phosphonium- and ammonium-based ionic liquids. J. Biotechnol. 2015, 206, 17-25.