5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride, 98%, Thermo Scientific Chemicals
5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride, 98%, Thermo Scientific Chemicals

A tricyclic serotnonin reuptake inhibitor
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number J65596.14
also known as J65596-14
Price (USD)/ Each
150.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
150.00
Add to cart
5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberJ65596.14
Price (USD)/ Each
150.00
-
Add to cart
Chemical Identifiers
CAS549-18-8
IUPAC Namehydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
Molecular FormulaC20H24ClN
InChI KeyKFYRPLNVJVHZGT-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2CCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (HPLC)98.0% min.
Loss on Drying6.0% max.
Melting Point195-200?C
FormPowder
A tricyclic serotnonin reuptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only