Thermo Scientific Chemicals
2'-Fluoro-2'-deoxycytidine, 99%
Catalog number: J65632.03
1 g, Each
Thermo Scientific Chemicals
2'-Fluoro-2'-deoxycytidine, 99%
Catalog number: J65632.03
1 g, Each
Quantity
Catalog number: J65632.03
also known as J65632-03
Price (USD)
Quantity
-
Have Questions?
Description
2'-Fluoro-2'-deoxycytidine is a potent inhibitor of the subgenomic hepatitis C virus replicon in Huh-7 cells. 2'-Fluoro-2'-deoxycytidine has been shown to inhibit Borna Disease virus replication and spread.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′-Fluoro-2′-deoxycytidine is a potent inhibitor of the subgenomic hepatitis C virus replicon in Huh-7 cells. 2′-Fluoro-2′-deoxycytidine has been shown to inhibit Borna Disease virus replication and spread.