GW 9662, Thermo Scientific Chemicals
GW 9662, Thermo Scientific Chemicals
Thermo Scientific Chemicals

GW 9662, Thermo Scientific Chemicals

An irreversible PPAR→ antagonist
Have Questions?
Quantity:
10 mg
Catalog number J65634.MA
also known as J65634-MA
Price (USD)/ Each
125.00
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
125.00
Add to cart
GW 9662, Thermo Scientific Chemicals
Catalog numberJ65634.MA
Price (USD)/ Each
125.00
-
Add to cart
Chemical Identifiers
CAS22978-25-2
IUPAC Name2-chloro-5-nitro-N-phenylbenzamide
Molecular FormulaC13H9ClN2O3
InChI KeyDNTSIBUQMRRYIU-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)C(=C1)C(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)White to off-white
Purity≥97.0%
An irreversible PPAR- antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only