Laminin A Chain (2091-2108)
Laminin A Chain (2091-2108)
Thermo Scientific Chemicals

Laminin A Chain (2091-2108)

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J65640.EXD
also known as J65640-!D
Price (USD)/ Each
152.00
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
152.00
Add to cart
Laminin A Chain (2091-2108)
Catalog numberJ65640.EXD
Price (USD)/ Each
152.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)2016.30
Product TypeLaminin A Chain (2091-2108)
Quantity0.5 mg
View more
Supports neurite outgrowth and stimulates neuronal-like process formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C