Thermo Scientific Chemicals

N-Acetyl-L-histidine monohydrate

Catalog number: J65657.06
5 g, Each
Thermo Scientific Chemicals

N-Acetyl-L-histidine monohydrate

Catalog number: J65657.06
5 g, Each
Quantity
Catalog number: J65657.06
also known as J65657-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39145-52-3
IUPAC Name
2-acetamido-3-(1H-imidazol-5-yl)propanoic acid hydrate
Molecular Formula
C8H13N3O4
InChI Key
PSWSDQRXCOJSFC-UHFFFAOYNA-N
SMILES
O.CC(=O)NC(CC1=CN=CN1)C(O)=O
Appearance (Color)
White to cream
Form
Powder or crystals or crystalline powder
Assay (Titration)
≥99.0 % (Calculated on anhydrous basis)
Assay (HPLC)
≥99.0 %
Optical Rotation
Specific Rotation: +44.0 to +49.0° (C=1, H₂O)

Description

N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for NÃŽ±-acetylhistidine synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for Nα-acetylhistidine synthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text