N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for NÃŽ±-acetylhistidine synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for Nα-acetylhistidine synthesis.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Serge N. Vinogradov. hydrogen bonds in crystal structures of amino acids, peptides and related molecules. International Journal of Peptide and Protein Research. 1979, 14, (4), 281-289.
Trevor G. Appleton.; Ferdinand J. Pesch.; Markus Wienken.; Stephan Menzer.; Bernhard Lippert. Linkage isomerism in square-planar complexes of platinum and palladium with histidine and derivatives. Inorg. Chem. 1992, 31, (21), 4410-4419.