Thermo Scientific Chemicals

Calcitonin Gene Related Peptide, Human

Catalog number: J65667.EXD
0.5 mg, Each
Thermo Scientific Chemicals

Calcitonin Gene Related Peptide, Human

Catalog number: J65667.EXD
0.5 mg, Each
Quantity
Catalog number: J65667.EXD
also known as J65667-!D
Price (USD)
Quantity
-
Have Questions?
Molecular Weight
Conforms
Assay (HPLC)
95.0% min.

Description

Calcitonin Gene Related Peptide is used as a potent, long-lasting vasodilator, activation of CGRP receptors on pancreatic β-cells increases plasma levels of pancreatic enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text