Calcitonin Gene Related Peptide, Human
Calcitonin Gene Related Peptide, Human
Thermo Scientific Chemicals

Calcitonin Gene Related Peptide, Human

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
0.5 mg
Catalog number J65667.MCR
also known as J65667-M^
Price (USD)/ Each
454.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
454.00
Add to cart
Calcitonin Gene Related Peptide, Human
Catalog numberJ65667.MCR
Price (USD)/ Each
454.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
SpeciesHuman
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)3789.3
Product TypeCalcitonin Gene Related Peptide, human
View more
Calcitonin Gene Related Peptide is used as a potent, long-lasting vasodilator, activation of CGRP receptors on pancreatic β-cells increases plasma levels of pancreatic enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C