Thermo Scientific Chemicals

Cytosine-beta-D-arabinofuranose hydrochloride

Catalog number: J65671.09
10 g, Each
Thermo Scientific Chemicals

Cytosine-beta-D-arabinofuranose hydrochloride

Catalog number: J65671.09
10 g, Each
Quantity
Catalog number: J65671.09
also known as J65671-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69-74-9
IUPAC Name
4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride
Molecular Formula
C9H14ClN3O5
InChI Key
KCURWTAZOZXKSJ-JBMRGDGGSA-N
SMILES
Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
Appearance (Color)
White to cream
Assay (HPLC)
≥ 99.0%
Loss on Drying
≤ 0.5%
Form
Powder or crystals or crystalline powder

Description

Interferes with the synthesis of DNA

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text